Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma Tubulin Antibody (TU-30), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB500574X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500574X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500574X Supplier Novus Biologicals Supplier No. NB500574AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

gamma Tubulin Monoclonal specifically detects gamma Tubulin in Human, Mouse, Rat, Porcine, Bovine, Chicken, Plant, Protozoa samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen gamma Tubulin
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone TU-30
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Gene Accession No. P23258
Gene Alias gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1, TUBG, TUBGCP1, tubulin gamma-1 chain, tubulin, gamma 1, tubulin, gamma polypeptide
Gene Symbols TUBG1
Host Species Mouse
Immunogen C-terminal peptide of gamma-tubulin counjugated to KLH.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Centrosome Markers, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 7283
Test Specificity The antibody TU-30 recognizes C-terminal peptide sequence EYHAATRPDYISWGTQ (aa 434-449) of gamma-tubulin, a 48 kDa structural constituent of cytoskeleton and microtubule organizing center (MTOC).
Target Species Human, Mouse, Rat, Pig, Bovine, Chicken, Plant, Protozoa
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.