Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma Tubulin Antibody (TU-32), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB190616549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB190616549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB190616549 Supplier Novus Biologicals Supplier No. NB11090616JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

gamma Tubulin Monoclonal specifically detects gamma Tubulin in Human, Porcine, Animal, Plant, Protozoa samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen gammaTubulin
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone TU-32
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1, TUBG, TUBGCP1, tubulin gamma-1 chain, tubulin, gamma 1, tubulin, gamma polypeptide
Gene Symbols TUBG1
Host Species Mouse
Immunogen human gamma-tubulin peptide EYHAATRPDYISWGTQ, amino acids 434-449
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cell Biology, Cellular Markers, Centrosome Markers, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 7283
Test Specificity The TU-32 recognizes an epitope (amino acids 434-449 in human) within C-terminus of gamma-tubulin, a 48 kDa structural constituent of cytoskeleton and microtubule organizing center (MTOC).
Target Species Multi-species, Plant, Pig, Protozoa
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.