Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell GAMMEX™ Non-Latex PI Surgical Gloves

Catalog No. 17010942 Shop All Ansell Products
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9

Polyisoprene, skin-friendly surgical glove with a soft glove formulation delivering extra comfort.

  • Optimal allergy protection: Being non-latex, these polyisoprene surgical gloves eliminate the risk of a latex allergy while minimizing sensitization to a Type IV chemical allergy as chemical accelerators known to cause skin allergies have been eliminated from its skin-friendly formulation
  • Superior comfort and dexterity: Ansell's proprietary soft yet strong formulation delivers exceptional fit and feel
  • Excellent grip and sensitivity: The smooth surface of these surgical gloves has a micro-textured finish to ensure confident handling and instrument grip during the most rigorous procedures
  • Effortless donning and doffing: A unique internal coating aids dry and damp hand donning, either when first donned or during intra-operative changes
  • Chemo-tested: Tested to ASTM D6978, these GAMMEX™ sterile gloves are safe to use with a wide range of chemotherapy drugs
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM D3577, ASTM D6978, ASTM D7160, ASTM F739, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, ISO 10282, JIS T9107
Certifications/Compliance (Europe) AS/NZS 4179, , EN 16523-1, EN 455 1-4
Accelerator Free No
Applications Healthcare
AQL 0.65
Color White
Disposable Single-use
Exterior Finish Smooth
Finger Thickness (English) 9.06 to 0.79 mil
Finger Thickness (Metric) 0.23 to 0.02 mm
Hand Specific
Latex Free Yes
Material Polyisoprene
Palm Thickness (English) 8.27 to 0.79 mil
Palm Thickness (Metric) 0.21 to 0.02 mm
Powdered Powder Free
Product Type Surgical Gloves
Size 5.5
Sterility Sterile
Thickness (English) 9.06 to 0.79 mil
Thickness (Metric) 0.23 to 0.02 mm
Packaging Type Dispenser Box
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.