Learn More
Description
Specifications
Specifications
| Antigen | Gankyrin |
| Applications | Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.97 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:500, Flow (Intracellular) 1:500 |
| Formulation | Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide |
| Gene Accession No. | O75832 |
| Gene Alias | 26S proteasome non-ATPase regulatory subunit 10, 26S proteasome regulatory subunit p28, ankyrin repeat protein, dJ889N15.2, gankyrin, hepatocellular carcinoma-associated protein p28-II, p28, p28(GANK), proteasome (prosome, macropain) 26S subunit, non-ATPase, 10 |
| Gene Symbols | PSMD10 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
