Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gankyrin Antibody (OTI3F6), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP271267A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP271267A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP271267A30 Supplier Novus Biologicals Supplier No. NBP271267AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Gankyrin Monoclonal specifically detects Gankyrin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen Gankyrin
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI3F6
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 26S proteasome non-ATPase regulatory subunit 10, 26S proteasome regulatory subunit p28, ankyrin repeat protein, dJ889N15.2, gankyrin, hepatocellular carcinoma-associated protein p28-II, p28, p28(GANK), proteasome (prosome, macropain) 26S subunit, non-ATPase, 10
Gene Symbols PSMD10
Host Species Mouse
Immunogen Full length recombinant protein of human Gankyrin (NP_002805) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Biology, Tumor Suppressors, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5716.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.