Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gankyrin Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB339815 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339815 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339815 Supplier Novus Biologicals Supplier No. NB1001834PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Gankyrin Polyclonal antibody specifically detects Gankyrin in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Flow (Intracellular)

Specifications

Antigen Gankyrin
Applications Western Blot, Immunocytochemistry, Flow Cytometry (Intracellular Staining)
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Flow (Intracellular)
Formulation PBS
Gene Alias 26S proteasome non-ATPase regulatory subunit 10, 26S proteasome regulatory subunit p28, ankyrin repeat protein, dJ889N15.2, gankyrin, hepatocellular carcinoma-associated protein p28-II, p28, p28(GANK), proteasome (prosome, macropain) 26S subunit, non-ATPase, 10
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human Gankyrin protein sequence (between residues 100-200). [UniProt# O75832]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Tumor Suppressors, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5716
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.