Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GANP Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | GANP |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-4331
![]() |
Bethyl Laboratories, Inc
A303127AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
GANP (germinal center-associated nuclear protein) is up-regulated selectively in germinal centers and carries an MCM-binding domain in the carboxyl-terminal side. It was found that a region (from 414 to 550 aa) in GANP molecule that is slightly similar to the known DNA-primase component p49. The recombit GANP fragment covering this region synthesizes RNA primers for extension by DNA polymerase I with single-stranded DNA templates in vitro. GANP DNA-primase activity is controlled by phosphorylation at Ser(502) that is induced by CD40-mediated signaling in vitro and in the germinal center B cells stimulated with antigen in vivo. Overexpression of ganp cDNA in Daudi B cells caused the increased DNA synthesis more than the levels of the mock-transfectants. These evidences suggested that the novel DNA-primase GANP is involved in regulation of cell proliferation of antigen-driven B cells in germinal centers.Specifications
GANP | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
GANP, MAP80, 80 kDa MCM3-associated protein, MCM3 import protein, MCM3 minichromosome maintece deficient 3 associated protein, germinal center-associated nuclear protein, minichromosome maintece deficient 3-associated protein | |
MCM3AP | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O60318 | |
8888 | |
Between 1 and 50 | |
Primary | |
4° C | |
MCM3AP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title