Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAP-43 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB362258 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362258 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362258 Supplier Novus Biologicals Supplier No. NB300143AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GAP-43 Polyclonal antibody specifically detects GAP-43 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Drosophila, Equine, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)

Specifications

Antigen GAP-43
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1
Host Species Rabbit
Immunogen C-terminal peptide of rodent GAP43, KEDPEADQEHA coupled to KLH.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2596
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.