Learn More
Description
Specifications
Specifications
| Antigen | GAP-43 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:400-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunohistochemistry-Paraffin reported in scientific literature (PMID 24686445), Immunohistochemistry-Frozen 1:400-1:1000, Immunoblotting reported in scientific literature (PMID 28112682) |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | P07936 |
| Gene Alias | Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1 |
| Gene Symbols | GAP43 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
