Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAP-43, p Ser41 Antibody, Novus Biologicals™
SDP

Catalog No. NB300179 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300179 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300179 Supplier Novus Biologicals Supplier No. NB300179
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GAP-43 Polyclonal specifically detects GAP-43 in Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen GAP-43 (p Ser41)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. P07936
Gene Alias Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1
Gene Symbols GAP43
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser41 conjugated to KLH. Accession # P07936
Molecular Weight of Antigen 50 kDa
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2596
Test Specificity Specific for the ∼50k Gap-43 protein phosphorylated at Ser41. In some tissues the antibody also recognizes a higher molecular weight protein that is also recognized by the pan Gap-43 antibody, that may be a Gap-43 aggregate or oligomer. Immunolableing is blocked by the phosphopeptide used as antigen but not by the corresponding dephosphopeptide. Immunolabeling is completely eliminated by treatment with gamma-Ptase.
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.