Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAPDH Antibody (13H12) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP227103SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227103SS 0.025 mg
NBP227103 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227103SS Supplier Novus Biologicals Supplier No. NBP227103SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 29 publications

GAPDH Monoclonal specifically detects GAPDH in Human, Mouse, Rat, Drosophila, Monkey, Primate, Sheep samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen GAPDH
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 13H12
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 0.25 - 1 ug/ml, Simple Western 1:25, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 1:10, Immunohistochemistry-Paraffin 5 ug/ml
Gene Accession No. P04406
Gene Alias aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH
Gene Symbols GAPDH
Host Species Mouse
Immunogen Amino acids between 275 and 325 of glyceraldehyde 3-phosphate dehydrogenase protein were used as the immunogen for this GAPDH antibody.
Molecular Weight of Antigen 36 kDa
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 2597
Test Specificity Reacts with Human, Primates/Monkey, Mouse and Drosophila. Based upon 91% sequence similarity with immunogen, this antibody is predicted to react with Guinea Pig, Sheep, Squirrel, Porcine/Pig, Ferret, Canine/Dog/Cat, Bovine, Reptile / Rattlesnake and several other species. Immunogen shows 82% similarity to Xenopus and Zebrafish.
Target Species Human, Mouse, Rat, Drosophila, Primate, Sheep
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.