Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAPDH Antibody (1D4), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 334 publications
$281.00 - $587.00
Specifications
Antigen | GAPDH |
---|---|
Clone | 1D4 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:1000, Simple Western 1:25, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation, Immunohistochemistry-Frozen, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Single Cell Western, KnockDown Validated |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB300221SS
![]() |
Novus Biologicals
NB300221SS |
0.025 mL |
Each for $281.00
|
|
|||||
NB300221
![]() |
Novus Biologicals
NB300221 |
0.1 mL |
Each for $587.00
|
|
|||||
Description
GAPDH Monoclonal specifically detects GAPDH in Human, Mouse, Rat, Porcine, Avian, Bovine, Canine, Chicken, Chinese Hamster, Drosophila, Equine, Feline, Hamster, Opossum, Primate, Rabbit, Sheep, Zebrafish samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Single Cell Western, Knockdown Validated.Specifications
GAPDH | |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control | |
50% PBS, 50% Glycerol with 5mM Sodium Azide | |
1D4 GAPDH, aging-associated gene 9 protein, Cytoplasm Marker, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, GAPDPeptidyl-cysteine S-nitrosylase GAPDH, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC102544, MGC102546, MGC103190, MGC103191, MGC105239, MGC127711, MGC88685, OCAS, p38, OCT1 coactivator in S phase, Peptidyl-cysteine S-nitrosylase GAPDH | |
GAPDH | |
IgM | |
Immunogen affinity purified | |
Human, mouse, rat, bovine, porcine, avian, sheep, opossum and primate. Likely to react with other mammals as well. Hamster and zebrafish reactivity reported by a customer review. Chicken reactivity reported in scientific literature (PMID: 24725420). |
1D4 | |
Western Blot 1:1000, Simple Western 1:25, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation, Immunohistochemistry-Frozen, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Single Cell Western, KnockDown Validated | |
Monoclonal | |
Supernatant | |
RUO | |
Human, Mouse, Rat, Pig, Avian, Bovine, Canine, Chicken, Chinese Hamster, Drosophila, Equine, Feline, Hamster, Opossum, Primate, Rabbit, Sheep, Zebrafish | |
P00355 | |
2597 | |
This GAPDH antibody was developed against full length purified porcine GAPDH [UniProt# P00355] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
36 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title