Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAPDH Antibody (CL3265), Novus Biologicals™
SDP

Catalog No. NB393163 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB393163 25 μL
NBP259025 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB393163 Supplier Novus Biologicals Supplier No. NBP25902525UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GAPDH Monoclonal specifically detects GAPDH in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen GAPDH
Applications Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown
Classification Monoclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000 - 1:10000, Knockdown Validated
Formulation PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide
Gene Alias aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH
Gene Symbols GAPDH
Host Species Mouse
Immunogen This GAPDH antibody was developed against a recombinant protein corresponding to the following amino acid sequence: TVKAENGKLVINGNPITIFQERDPSKIKWGDAGAEYVVESTGVFTTMEKAGAHLQGGAKRVIIS
Molecular Weight of Antigen 36 kDa
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 2597
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.