Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAPDH Antibody, Novus Biologicals™
SDP

Catalog No. NB300327 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300327 0.1 mL
NB018158 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300327 Supplier Novus Biologicals Supplier No. NB300327
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 33 publications

GAPDH Polyclonal specifically detects GAPDH in Human, Mouse, Rat, Porcine, Bacteria, Bovine, Chicken, Equine, Fungi, Invertebrate, Yeast samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GAPDH
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:5000, Simple Western 1:500, Immunohistochemistry 1:10000, Immunocytochemistry/Immunofluorescence 1:500-1:1000
Formulation Supplied as serum with 5mM Sodium Azide
Gene Accession No. P10096
Gene Alias aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH
Gene Symbols GAPDH
Host Species Rabbit
Immunogen This GAPDH antibody was developed against full length recombinant human GAPDH
Molecular Weight of Antigen 36 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 2597
Test Specificity Human, cow, pig, mouse, rat, bird and chicken. Yeast reactivity reported in scientific literature (PMID: 23709183). Since GAPDH is one of the most conserved proteins known and it is therefore likely that the antibody will react with other species also. Invertebrate reactivity reported in scientific literature (PMID: 18638509).
Target Species Human, Mouse, Rat, Pig, Bacteria, Bovine, Chicken, Equine, Fungi, Invertebrate, Yeast
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.