Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAPDH Mouse anti-Human, Mouse, Rat, Porcine, Amphibian, Canine, Chinese Hamster, Feline, Fish, Primate, Monkey, Rabbit, Bovine (Negative), Goat (Negative), Clone: 6C5cc, Novus Biologicals™
SDP

Catalog No. NB141861 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB141861 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB141861 Supplier Novus Biologicals Supplier No. NB6005020.2MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 57 publications

GAPDH Monoclonal specifically detects GAPDH in Human, Mouse, Rat, Porcine, Amphibian, Canine, Chinese Hamster, Feline, Fish, Primate, Monkey, Rabbit, Bovine (Negative), Goat (Negative) samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunoassay.

Specifications

Antigen GAPDH
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunoassay
Classification Monoclonal
Clone 6C5cc
Conjugate Unconjugated
Dilution Western Blot 0.5-1 ug/ml, Simple Western 1:100, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation, Immunoassay
Formulation PBS (pH 7.4)
Gene Accession No. P04406
Gene Alias aging-associated gene 9 protein, anti-GAPDH 6c5, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, gapdh 6c5, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH
Host Species Mouse
Immunogen This GAPDH antibody was developed against human or rabbit GAPDH.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 2597
Target Species Human, Mouse, Rat, Pig, Amphibian, Canine, Chinese Hamster, Feline, Fish, Primate, Monkey, Rabbit, Bovine (Negative), Goat (Negative)
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.