Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GARNL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18798225UL
Description
GARNL1 Polyclonal specifically detects GARNL1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GARNL1 | |
Polyclonal | |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
DKFZp566D133, DKFZp667F074, GAP-related interacting protein to E12, GARNL1p240, GRIPEGAP-related-interacting partner to E12, GTPase activating RANGAP domain-like 1, GTPase activating Rap/RanGAP domain-like 1, KIAA0884GTPase-activating Rap/Ran-GAP domain-like 1, Ral GTPase activating protein, alpha subunit 1 (catalytic), ral GTPase-activating protein alpha subunit 1, ral GTPase-activating protein subunit alpha-1, RalGAPalpha1, Tuberin-like protein 1, TULIP1Tulip1 | |
Rabbit | |
Affinity Purified | |
RUO | |
253959 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
RALGAPA1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LVSCIQIRSEENMPGGGLSAGLASANSNVRIIVRDLSGKYSWDSAILYGPPPVSGLSEPTSFMLSLSHQEKPEEPPTSNECLEDITVKDGLSLQFKRFRETVPTWDTIRDEEDVLDELLQYLGVTSPECLQRTGISLNIPAPQPVCISE | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction