Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GARNL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | GARNL1 |
---|---|
Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
GARNL1 Polyclonal specifically detects GARNL1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GARNL1 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
253959 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LVSCIQIRSEENMPGGGLSAGLASANSNVRIIVRDLSGKYSWDSAILYGPPPVSGLSEPTSFMLSLSHQEKPEEPPTSNECLEDITVKDGLSLQFKRFRETVPTWDTIRDEEDVLDELLQYLGVTSPECLQRTGISLNIPAPQPVCISE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
Polyclonal | |
Rabbit | |
Human | |
DKFZp566D133, DKFZp667F074, GAP-related interacting protein to E12, GARNL1p240, GRIPEGAP-related-interacting partner to E12, GTPase activating RANGAP domain-like 1, GTPase activating Rap/RanGAP domain-like 1, KIAA0884GTPase-activating Rap/Ran-GAP domain-like 1, Ral GTPase activating protein, alpha subunit 1 (catalytic), ral GTPase-activating protein alpha subunit 1, ral GTPase-activating protein subunit alpha-1, RalGAPalpha1, Tuberin-like protein 1, TULIP1Tulip1 | |
RALGAPA1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title