Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GARP Monoclonal Antibody (YGIC86), APC, eBioscience™
GREENER_CHOICE

Catalog No. p-7173274
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-153-46 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-153-46 Supplier Invitrogen™ Supplier No. 17989182
Only null left
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The YGIC86 monoclonal antibody reacts with mouse Glycoprotein A Repetitions Predominant (GARP, also known as Lrrc32 or Garpin). GARP is an approximately 80 kDa glycoprotein that is expressed on the cell surface. Using northern blot, RT-PCR or microarray analyses, the expression of GARP has been reported in placenta, lung, kidney, heart, ovary, liver, skeletal muscle, and pancreas. Protein expression has been observed on megakaryocytes, platelets and activated regulatory T (Treg) cells. The expression of GARP on Treg cells has been reported to be necessary for the suppressive function of Treg cells, possibly related to its role as a cell surface receptor for LAP/TGF beta. When looking for expression of GARP on platelets, it is recommended to use the Foxp3 Staining Buffer Set (Product # 00-5523) to fix the platelets before staining with this YGIC86 monoclonal antibody. Fixation/permeabilization is not necessary when staining Treg cells. YGIC86 does not cross-react to rat GARP. Applications Reported: This YGIC86 antibody has been reported for use in flow cytometric analysis. Applications Tested: This YGIC86 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is rec...

GARP, also known as leucine rich repeat containing 32, is a type I membrane protein with twenty-two leucine-rich repeats and a short intracytoplasmic region. GARP was identified based on the analysis of gene expression profiling of Treg and Th cells following TCR stimulation. Since GARP is expressed in CD4+, CD25+, FoxP3+ cells (Tregs), it may be a candidate for cell surface marker for Treg cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GARP
Applications Flow Cytometry
Classification Monoclonal
Clone YGIC86
Concentration 0.2 mg/mL
Conjugate APC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene LRRC32
Gene Accession No. G3XA59
Gene Alias AI426318; D11S833E; D11S833Eh; D7H11S833E; EG434215; Garp; garpin; Glycoprotein A repetitions predominant; leucine rich repeat containing 32; leucine-rich repeat-containing protein 32; Lrrc32; Transforming growth factor beta activator LRRC32
Gene Symbols LRRC32
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 434215
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.