Learn More
GARP Rabbit anti-Human, Rat, Polyclonal, FabGennix
Description
Specifications
Specifications
| Antigen | GARP |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5-1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 |
| Gene | LRRC32 |
| Gene Accession No. | Q14392 |
| Gene Alias | AI426318; D11S833E; D11S833Eh; D7H11S833E; EG434215; Garp; garpin; Glycoprotein A repetitions predominant; leucine rich repeat containing 32; leucine-rich repeat-containing protein 32; Lrrc32; Transforming growth factor beta activator LRRC32 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.