Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GARS Antibody (4E10), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP242679A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP242679A70 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP242679A70 Supplier Novus Biologicals Supplier No. NBP242679AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GARS Monoclonal specifically detects GARS in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GARS
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 40000000000
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias AP-4-A synthetase, Charcot-Marie-Tooth neuropathy 2D, Charcot-Marie-Tooth neuropathy, neuronal type, D, Diadenosine tetraphosphate synthetase, DSMAV, glycine tRNA ligase, Glycine--tRNA ligase, glycyl-tRNA synthetase, GlyRSEC 6.1.1.14, HMN5, SMAD1CMT2D
Gene Symbols GARS
Host Species Mouse
Immunogen Recombinant human GARS (43-289aa) purified from E. coli
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 2617
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.