Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GARS Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | GARS |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
GARS Polyclonal specifically detects GARS in Human samples. It is validated for Western Blot.Specifications
| GARS | |
| Polyclonal | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| AP-4-A synthetase, Charcot-Marie-Tooth neuropathy 2D, Charcot-Marie-Tooth neuropathy, neuronal type, D, Diadenosine tetraphosphate synthetase, DSMAV, glycine tRNA ligase, Glycine--tRNA ligase, glycyl-tRNA synthetase, GlyRSEC 6.1.1.14, HMN5, SMAD1CMT2D | |
| GARS | |
| IgG | |
| 83 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| P41250 | |
| 2617 | |
| Synthetic peptides corresponding to GARS(glycyl-tRNA synthetase) The peptide sequence was selected from the middle region of GARS. Peptide sequence IEPSFGLGRIMYTVFEHTFHVREGDEQRTFFSFPAVVAPFKCSVLPLSQN. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title