Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Gas Anesthesia System
SDP

Stoelting™ Complete Anesthesia System was designed specifically for stereotaxic surgery. Includes a tabletop and/or mobile cart with Isoflurane Vaporizer, Active Gas Scavenger and continuous flow Induction Chamber.

$16120.00 - $17880.00

Specifications

For Use With (Equipment) Stereotaxic surgery
View More Specs

Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
10-000-939
SDP
View Documents
Stoelting™
53940
Tabletop w/ Induction Chamber
Each for $16,120.00
Only null left
Add to Cart
 
10-000-940
SDP
View Documents
Stoelting™
53941
Tabletop/Mobile w/Induction Chamber
Each for $17,880.00
Only null left
Add to Cart
 
Description

Description

Inhalant Anesthesia System for Stereotaxic Surgery with Active Gas Waste Removal inlcudes:

  • Tabletop and/or Mobile Anesthesia System Complete with Isoflurane Vaporizer
  • Active Gas Scavenger
  • Continuous Induction Chamber

Features:

  • No flowmeters to adjust. Flip a toggle switch to on to start flow of fresh gas, indicator confirms flow
  • 3 stations for anesthesia with preset flow rate for any station (500cc-5 LPM)
  • Color coded tubing and connector to help user identify which fresh gas supply tubing goes to which station
  • Pressure gauge to let user know when pressure has been released from system for filling of vaporizer.
  • Generates a 50 LPM negative flow that can be attenuated to appropriate levels and divided between 3 stations.
  • Direct vacuum port for use with Vapor-Vac Hooded Induction Chamber
    Adjustable negative flow meters easily activated with toggle switches.
  • Waste anesthetic gases can be discharged into activated charcoal canister or any non re-circulating hood, vent, downdraft/ back draft system or snorkel.
  • Available without negative flow generating pump- can then be used to attenuate active, in-house vacuum source.
  • Designed for use with the Posi-vac Non-Rebreathing system, Vapor-Vac hooded induction chamber, Stereotaxic nose cone or SCOOP, 3EX Vaporizer. (not included)
  • Higher Throughput
  • Intuitive system- easy to operate, precise, safe flow rate for fresh gas
    Healthy for subjects- less morbidity, less mortality
Specifications

Specifications

Stereotaxic surgery
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.