Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gas1 Antibody (11C47), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB222032488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB222032488 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB222032488 Supplier Novus Biologicals Supplier No. NBP222032AF488
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Gas1 Monoclonal specifically detects Gas1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Gas1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 11C47
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias GAS-1, growth arrest-specific 1, Growth arrest-specific gene-1, growth arrest-specific protein 1
Gene Symbols GAS1
Host Species Rat
Immunogen Human recombinant Gas1
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2619
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.