Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gas1 Antibody (11C47) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP2220320.025MG
Description
Gas1 Monoclonal specifically detects Gas1 in Human samples. It is validated for Western Blot, ELISA.Specifications
Gas1 | |
Monoclonal | |
LYOPH | |
Western Blot 1:500-1:1000, ELISA 1:2000 | |
NP_002039 | |
GAS1 | |
This antibody was produced from a hybridoma (rat myeloma fused with spleen cells from a rat immunized with purified recombinant human Gas1. Rat anti human Gas1 NP_002039. | |
0.025 mg | |
Primary | |
Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2 |
ELISA, Western Blot | |
11C47 | |
Unconjugated | |
A 0.2 um filtered solution in PBS with No Preservative | |
GAS-1, growth arrest-specific 1, Growth arrest-specific gene-1, growth arrest-specific protein 1 | |
Rat | |
Protein G purified | |
RUO | |
2619 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction