Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gas1 Antibody (11C47) - Azide and BSA Free, Novus Biologicals™
SDP

Rat Monoclonal Antibody

$206.50 - $560.50

Specifications

Antigen Gas1
Clone 11C47
Concentration LYOPH
Applications ELISA, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB019293
SDP
View Documents
Novus Biologicals
NBP2220320.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP222032
SDP
View Documents
Novus Biologicals
NBP222032
0.1 mg
Each for $560.50
Only null left
Add to Cart
 
Description

Description

Gas1 Monoclonal specifically detects Gas1 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Gas1
LYOPH
Monoclonal
Purified
RUO
NP_002039
2619
IgG2
Protein G purified
11C47
ELISA, Western Blot
Unconjugated
Rat
Human
GAS-1, growth arrest-specific 1, Growth arrest-specific gene-1, growth arrest-specific protein 1
GAS1
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.