Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAS2 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB055615 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB055615 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB055615 Supplier Novus Biologicals Supplier No. NBP298799DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GAS2 Polyclonal specifically detects GAS2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GAS2
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias GAS-2, growth arrest-specific 2, growth arrest-specific protein 2, MGC32610
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human GAS2.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 2620
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.