Learn More
Description
Specifications
Specifications
| Antigen | GAS2L3 |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | FITC |
| Dilution | Immunoprecipitation, Western Blot (Negative) |
| Formulation | PBS |
| Gene Alias | GAS2-like protein 3, growth arrest-specific 2 like 3, Growth arrest-specific protein 2-like 3 |
| Host Species | Rabbit |
| Immunogen | Maps to a region between residue 325 to 375 of human Growth Arrest-Specific 2 Like 3 using the numbering given in entry NP_777602.1 (GeneID 283431). |
| Purification Method | Immunogen affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
