Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAS2L3 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB290179 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290179 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290179 Supplier Novus Biologicals Supplier No. NBP230358F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GAS2L3 Polyclonal antibody specifically detects GAS2L3 in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative)

Specifications

Antigen GAS2L3
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation (Negative)
Formulation PBS
Gene Alias GAS2-like protein 3, growth arrest-specific 2 like 3, Growth arrest-specific protein 2-like 3
Host Species Rabbit
Immunogen Maps to a region between residue 1 to 50 of human Growth Arrest-Specific 2 Like 3 using the numbering given in entry NP_777602.1 (GeneID 283431).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 283431
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.