Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gas6 Antibody (RM0084-6J2), PerCP, Novus Biologicals™
SDP

Catalog No. NB362030 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362030 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362030 Supplier Novus Biologicals Supplier No. NBP121724PCP
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Gas6 Monoclonal antibody specifically detects Gas6 in Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen Gas6
Applications Western Blot, ELISA
Classification Monoclonal
Clone RM0084-6J2
Conjugate PerCP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6
Host Species Rat
Immunogen Mouse recombinant Gas6
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2621
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.