Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAS8 Antibody (OTI10G2), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP27079A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27079A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27079A94 Supplier Novus Biologicals Supplier No. NBP270795AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GAS8 Monoclonal specifically detects GAS8 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen GAS8
Applications Western Blot
Classification Monoclonal
Clone OTI10G2
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias GAS-11, GAS11MGC138326, GAS-8, growth arrest specific 11, growth arrest-specific 11, growth arrest-specific 8, Growth arrest-specific protein 11, growth arrest-specific protein 8
Gene Symbols GAS8
Host Species Mouse
Immunogen Full length recombinant protein of human GAS8 (NP_001472) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2622
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.