Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gastric Lipase Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB339093 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339093 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339093 Supplier Novus Biologicals Supplier No. NB11060930MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Gastric Lipase Polyclonal antibody specifically detects Gastric Lipase in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen Gastric Lipase
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 3.1.1, EC 3.1.1.3, Gastric lipase, gastric triacylglycerol lipase, GL, HGL, HLAL, lipase, gastric, MGC138477, MGC142271
Host Species Rabbit
Immunogen A synthetic peptide within residues 1-100 of human Gastric Lipase. [Swiss-Prot# P07098]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Endothelial Cell Markers, Glia Markers, Growth and Development, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8513
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.