Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gastrin Antibody (GAST/2632), HRP, Novus Biologicals™
SDP

Catalog No. NB379253 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379253 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379253 Supplier Novus Biologicals Supplier No. NBP308658H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Gastrin Monoclonal antibody specifically detects Gastrin in Human samples. It is validated for ELISA

Specifications

Antigen Gastrin
Applications ELISA
Classification Monoclonal
Clone GAST/2632
Conjugate HRP
Formulation PBS
Gene Alias GAS, gastrin
Host Species Mouse
Immunogen Recombinant full-length human Gastrin protein (Uniprot: P01350)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cell Biology, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2520
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.