Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gastrin Antibody, Novus Biologicals™
SDP

Catalog No. NB036595 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036595 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036595 Supplier Novus Biologicals Supplier No. NBP289004
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Gastrin Polyclonal specifically detects Gastrin in Human, Rat, Porcine samples. It is validated for Immunohistochemistry.

Specifications

Antigen Gastrin
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Neat serum
Gene Alias GAS, gastrin
Host Species Rabbit
Immunogen Synthetic peptide corresponding to aa 18-34 of Gastrin (gastrin-17; gastrin-34).
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2520
Target Species Human, Rat, Pig
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.