Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gastrin-releasing Peptide/Bombesin/Neuromedin C Antibody, Novus Biologicals™
SDP

Catalog No. NB036596 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036596 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036596 Supplier Novus Biologicals Supplier No. NBP289005
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Gastrin-releasing Peptide/Bombesin/Neuromedin C Polyclonal specifically detects Gastrin-releasing Peptide/Bombesin/Neuromedin C in Non-species specific samples. It is validated for Immunohistochemistry-Frozen.

Specifications

Antigen Gastrin-releasing Peptide/Bombesin/Neuromedin C
Applications Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Formulation Neat serum
Gene Alias BN, bombesin, gastrin-releasing peptide, GRP-10, neuromedin C, pre-progastrin releasing peptide, preproGRP, proGRP
Host Species Rabbit
Immunogen Synthetic Gastrin-releasing Peptide.
Quantity 50 μL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2922
Target Species Non-species specific
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.