Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gastrin-releasing Peptide R/GRPR Antibody, Novus Biologicals™
SDP

Catalog No. NBP146576 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146576 0.1 mL
NB018628 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP146576 Supplier Novus Biologicals Supplier No. NBP146576
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Gastrin-releasing Peptide R/GRPR Polyclonal specifically detects Gastrin-releasing Peptide R/GRPR in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Gastrin-releasing Peptide R/GRPR
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:200-1:2000, Immunohistochemistry 1:200-1:2000, Immunohistochemistry-Paraffin 1:200-1:2000
Formulation Lyophilized from whole antisera with No Preservative
Gene Accession No. P30550, P21729, P52500
Gene Alias gastrin-releasing peptide receptor, GRP-preferring bombesin receptor, GRP-R
Gene Symbols GRPR
Host Species Rabbit
Immunogen A synthetic peptide from human Gastrin-releasing Peptide R/GRPR conjugated to blue carrier protein was used as the antigen.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2925
Test Specificity Specific for GRPR.
Reconstitution Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.