Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA-1 Antibody (4F5), Novus Biologicals™
SDP

Catalog No. NBP147492 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147492 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147492 Supplier Novus Biologicals Supplier No. NBP147492
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

GATA-1 Monoclonal specifically detects GATA-1 in Human, Baboon samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen GATA-1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4F5
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Chromatin Immunoprecipitation (ChIP)
Gene Alias Eryf1, ERYF1GATA-binding protein 1 (globin transcription factor 1), GATA binding protein 1 (globin transcription factor 1), GATA-1erythroid transcription factor, GATA-binding factor 1, GF-1, GF1globin transcription factor 1, NF-E1 DNA-binding protein, NFE1erythroid transcription factor 1, transcription factor GATA1, XLTT
Gene Symbols GATA1
Host Species Mouse
Immunogen Purified recombinant full length human GATA1 expressed in E. Coli.
Molecular Weight of Antigen 42.7 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2623
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.