Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA-3 Antibody (GATA3/2445), HRP, Novus Biologicals™
SDP

Catalog No. NB392318 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392318 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392318 Supplier Novus Biologicals Supplier No. NBP308425H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GATA-3 Monoclonal antibody specifically detects GATA-3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Protein Array

Specifications

Antigen GATA-3
Applications Western Blot, Flow Cytometry, ELISA, Protein Array
Classification Monoclonal
Clone GATA3/2445
Conjugate HRP
Formulation PBS
Gene Alias 131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3
Host Species Mouse
Immunogen Recombinant fragment of human GATA-3 protein (around aa 357-436) (exact sequence is proprietary) (Uniprot: P23771)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 2625
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.