Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA-3 Antibody (OTI8H4), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP270796UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270796UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270796UV Supplier Novus Biologicals Supplier No. NBP270796UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GATA-3 Monoclonal specifically detects GATA-3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen GATA-3
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI8H4
Conjugate DyLight 350
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3
Gene Symbols GATA3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 155-443 of human GATA-3 (NP_002042) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 2625.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.