Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA-4 Antibody (6H10), Novus Biologicals™
SDP

Catalog No. NBP147540 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147540 0.1 mL
NBP147540SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP147540 Supplier Novus Biologicals Supplier No. NBP147540
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 1 publication

GATA-4 Monoclonal specifically detects GATA-4 in Human, Mouse, Rat samples. It is validated for ELISA, Western Blot.

Specifications

Antigen GATA-4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6H10
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000
Formulation Ascites with 0.03% Sodium Azide
Gene Accession No. P43694
Gene Alias GATA binding protein 4, GATA-binding factor 4, GATA-binding protein 4, MGC126629, transcription factor GATA-4
Gene Symbols GATA4
Host Species Mouse
Immunogen Purified recombinant fragment of human GATA4 expressed in E. coli. [UniProt# P43694]
Molecular Weight of Antigen 46 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2626
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.