Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA-4 Antibody (6H10), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 1 publication

$181.90 - $487.50

Specifications

Antigen GATA-4
Clone 6H10
Dilution Western Blot 1:500-1:2000, ELISA 1:10000
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP147540SS
SDP
View Documents
Novus Biologicals
NBP147540SS
0.025 mL
Each for $181.90
Only null left
Add to Cart
 
NBP147540
SDP
View Documents
Novus Biologicals
NBP147540
0.1 mL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

GATA-4 Monoclonal specifically detects GATA-4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

GATA-4
Western Blot 1:500-1:2000, ELISA 1:10000
Monoclonal
Ascites
RUO
Human, Mouse, Rat
P43694
2626
Purified recombinant fragment of human GATA4 expressed in E. coli. [UniProt# P43694]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
6H10
Western Blot, ELISA
Unconjugated
Mouse
Mesenchymal Stem Cell Markers, Stem Cell Markers
Ascites with 0.03% Sodium Azide
GATA binding protein 4, GATA-binding factor 4, GATA-binding protein 4, MGC126629, transcription factor GATA-4
GATA4
IgG2b
Unpurified
46 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.