Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA-4 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP224585SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224585SS 0.025 mg
NBP224585 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224585SS Supplier Novus Biologicals Supplier No. NBP224585SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

GATA-4 Polyclonal specifically detects GATA-4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GATA-4
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 10 ug/ml. Use reported by customer review
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P43694
Gene Alias GATA binding protein 4, GATA-binding factor 4, GATA-binding protein 4, MGC126629, transcription factor GATA-4
Gene Symbols GATA4
Host Species Rabbit
Immunogen A portion of amino acids 330-380 of human GATA-4 was used as the immunogen for the antibody.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2626
Test Specificity The amino acid sequence used as immunogen for the HSP27 antibody is 100% homologous in human, horse, macaque, chimpanzee and dog, 90% homologous in opossum, cow, zebrafish, boar and Xenopus and 81% homologous in rat and mouse.
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.