Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA-4 Antibody (OTI9F9), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP270797A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP270797A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP270797A30 Supplier Novus Biologicals Supplier No. NBP270797AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GATA-4 Monoclonal specifically detects GATA-4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GATA-4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI9F9
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias GATA binding protein 4, GATA-binding factor 4, GATA-binding protein 4, MGC126629, transcription factor GATA-4
Gene Symbols GATA4
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 296-442 of human GATA-4 (NP_002043) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2626
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.