Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GATA-6 Antibody (SR2056), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321806100UL
Description
GATA-6 Monoclonal antibody specifically detects GATA-6 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
GATA-6 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
SR2056 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
GATA binding protein 6, GATA-binding factor 6, GATA-binding protein 6, transcription factor GATA-6 | |
A synthesized peptide derived from human GATA-6 (Uniprot #: Q92908) | |
100 μg | |
Phospho Specific, Stem Cell Markers | |
2627 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction