Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GATA-6 Antibody (SR2056), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | GATA-6 |
---|---|
Clone | SR2056 |
Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Description
GATA-6 Monoclonal antibody specifically detects GATA-6 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
GATA-6 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
GATA binding protein 6, GATA-binding factor 6, GATA-binding protein 6, transcription factor GATA-6 | |
A synthesized peptide derived from human GATA-6 (Uniprot #: Q92908) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2056 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Phospho Specific, Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
2627 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title