Learn More
GATA3 Mouse, Alexa Fluor 488, Clone: L50-823, BD
Mouse Monoclonal Antibody
$327.50 - $484.00
Specifications
Antigen | GATA3 |
---|---|
Clone | L50-823 |
Classification | Monoclonal |
Conjugate | Alexa Fluor 488 |
Host Species | Mouse |
Description
GATA3 (GATA binding protein 3) is a member of the GATA family of transcription factors. This ∽50-kDa nuclear protein regulates the development and subsequent maintenance of multiple tissues. GATA3 is involved in the development of T lymphocytes (regulates T cell receptor subunit gene expression) and the differentiation of mature T cells to become Th2 cells. The expressed levels of normal or mutant GATA3 are also associated with the behaviors of various cancer cells including estrogen receptor-positive breast carcinoma cells.
The L50-823 monoclonal antibody recognizes human and mouse GATA3.
Specifications
GATA3 | |
Monoclonal | |
Mouse | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified |
L50-823 | |
Alexa Fluor 488 | |
RUO | |
Conserved peptide between the trans-activation and DNA-binding domains of human, mouse and rat GATA3 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.