Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATA3 Oligo, Mouse Anti-Human, Clone: L50-823, BD AbSeq™

Catalog No. BDB570750 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570750 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB570750 Supplier BD Biosciences Supplier No. 940521
Only null left
Add to Cart
Add to Cart

Mouse Oligoclonal Antibody

GATA3 Oligo Oligoclonal antibody specifically detects GATA3 Oligo in samples. It is validated for Single Cell 3'Sequencing
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATA3 Oligo
Applications Single Cell 3' Sequencing
Classification Oligoclonal
Clone L50-823
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias Gata-3; GATA binding protein 3; GATA-binding factor 3; HDR
Host Species Mouse
Immunogen Conserved peptide between the trans-activation and DNA-binding domains of human, mouse and rat GATA3
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2625, 14462
Target Species None
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.