Learn More
GATA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 104171AP150UL

Description
This is a rabbit polyclonal antibody raised against part of C-terminus of human GATA3.
Trans-acting T-cell specific transcription factor GATA-3 is a member of GATA family of transcription factors that regulates development of multiple tissues. It is an important transcription factor in regulating human Th2 cell differentiation in vivo.Specifications
GATA3 | |
Polyclonal | |
Unconjugated | |
GATA3 | |
GATA binding factor 3, GATA binding protein 3, GATA3, HDR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14462, 2625, 85471 | |
-20°C | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P23771, P23772 | |
Gata3 | |
GATA3 Fusion Protein Ag0174 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.