Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GATA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | GATA3 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6291
![]() ![]() |
Proteintech Group Inc
104171AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This is a rabbit polyclonal antibody raised against part of C-terminus of human GATA3.
Trans-acting T-cell specific transcription factor GATA-3 is a member of GATA family of transcription factors that regulates development of multiple tissues. It is an important transcription factor in regulating human Th2 cell differentiation in vivo.Specifications
GATA3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P23771, P23772 | |
14462, 2625, 85471 | |
GATA3 Fusion Protein Ag0174 | |
Primary | |
-20°C | |
GATA3 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GATA binding factor 3, GATA binding protein 3, GATA3, HDR | |
Gata3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title