Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATE-16/GABARAPL2 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB185037 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB185037 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB185037 Supplier Novus Biologicals Supplier No. NBP282090G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GATE-16/GABARAPL2 Polyclonal antibody specifically detects GATE-16/GABARAPL2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen GATE-16/GABARAPL2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias ATG8, ATG8C, FLC3A, gamma-aminobutyric acid receptor-associated protein-like 2, Ganglioside expression factor 2, GATE16, GATE-16, GEF2, GEF-2, GEF2GEF-2, General Protein Transport Factor P16, Golgi-Associated ATPase Enhancer Of 16 KDa, MAP1 light chain 3 related protein, MAP1 light chain 3-related protein
Host Species Rabbit
Immunogen GABARAPL2 antibody was raised against a 15 amino acid peptide near the carboxy terminus of human GABARAPL2. The immunogen is located within the last 50 amino acids of GABARAPL2. Amino Acid Squence: MGQLYEKEKDEDGFLY
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Golgi Apparatus Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 11345
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.