Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GATE-16/GABARAPL2 Rabbit anti-Human, Mouse, Rat, Clone: 8Z8M8, Novus Biologicals™
SDP

Catalog No. NB168753 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168753 20 μg
NB168752 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168753 Supplier Novus Biologicals Supplier No. NBP31683220UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GATE-16/GABARAPL2 Monoclonal antibody specifically detects GATE-16/GABARAPL2 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen GATE-16/GABARAPL2
Applications Western Blot
Classification Monoclonal
Clone 8Z8M8
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ATG8, ATG8C, FLC3A, gamma-aminobutyric acid receptor-associated protein-like 2, Ganglioside expression factor 2, GATE16, GATE-16, GEF2, GEF-2, GEF2GEF-2, General Protein Transport Factor P16, Golgi-Associated ATPase Enhancer Of 16 KDa, MAP1 light chain 3 related protein, MAP1 light chain 3-related protein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 38-117 of human GATE-16/GABARAPL2 (P60520).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Autophagy, Golgi Apparatus Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 11345
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.